Sulfuric acid, 20% fuming, 18-24% free SO{3}, Thermo Scientific Chemicals
Sulfuric acid, 20% fuming, 18-24% free SO{3}, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfuric acid, 20% fuming, 18-24% free SO{3}, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
4 x 500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045543.22
also known as 045543-22
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
59.50 
Save 8.85 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
59.50 
Save 8.85 (15%)
Add to cart
Sulfuric acid, 20% fuming, 18-24% free SO{3}, Thermo Scientific Chemicals
Catalog number045543.22
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
59.50 
Save 8.85 (15%)
-
Add to cart
Chemical Identifiers
CAS8014-95-7
IUPAC Namesulfonylideneoxidane; sulfuric acid
Molecular FormulaH2O7S2
InChI KeyHIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILESO=S(=O)=O.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Suppliers CofA≥104.05 to ≤105.4% (w/w)
Assay from Suppliers CofAFree SO₃: ≥18 to ≤24% (w/w)
Impurity contentNitrate, NO₃: Reported
Appearance (Color)Cloudy white
View more
Sulfuric acid is a strong dehydrating agent and is applied in many applications like domestic acidic drain cleaners, electrolytes in lead-acid batteries and various cleansing agents. It is useful for mineral processing, fertilizer manufacture, petroleum purification, wastewater processing and chemical synthesis. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows a useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only