Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, Thermo Scientific Chemicals
Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
500 mg
Catalog number 045565.04
also known as 045565-04
Price (USD)/ Each
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS12354-84-6
IUPAC Namediiridium(3+) bis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Molecular FormulaC20H30Cl4Ir2
InChI KeyMMAGMBCAIFVRGJ-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3].C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.0%
Appearance (Color)Orange
FormCrystals or crystalline powder
Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer is used as a precursor to catalysts for the asymmetric transfer hydrogenation of ketones. It is a catalyst for greener amine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only