Thermo Scientific Chemicals

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, Thermo Scientific Chemicals

Catalog number: 045565.ME
500 mg, Each
Thermo Scientific Chemicals

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, Thermo Scientific Chemicals

Catalog number: 045565.ME
500 mg, Each
Quantity
Catalog number: 045565.ME
also known as 045565-ME
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12354-84-6
IUPAC Name
diiridium(3+) bis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Molecular Formula
C20H30Cl4Ir2
InChI Key
MMAGMBCAIFVRGJ-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3].C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
Assay from Supplier's CofA
≥97.0%
Appearance (Color)
Orange
Form
Crystals or crystalline powder

Description

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer is used as a precursor to catalysts for the asymmetric transfer hydrogenation of ketones. It is a catalyst for greener amine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text