Thermo Scientific Chemicals
Platinum 30 wt% Rhodium wire, 0.33mm (0.013in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number: 045571.G2
2 m, Each
Thermo Scientific Chemicals
Platinum 30 wt% Rhodium wire, 0.33mm (0.013in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Catalog number: 045571.G2
2 m, Each
Quantity
Catalog number: 045571.G2
also known as 045571-G2