Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals
Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals

Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.
Have Questions?
Quantity:
1 Pc.
Catalog number 045575.KG
also known as 045575-KG
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
1 Pc.
Request bulk or custom format
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
Add to cart
Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number045575.KG
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10043-11-5
IUPAC Nameboranimine
Molecular FormulaBN
InChI KeyLNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILESB=N
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only