Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water, Thermo Scientific Chemicals
Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 045576.09
also known as 045576-09
Price (USD)/ Each
670.65
Online exclusive
745.00 
Save 74.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
670.65
Online exclusive
745.00 
Save 74.35 (10%)
Add to cart
Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water, Thermo Scientific Chemicals
Catalog number045576.09
Price (USD)/ Each
670.65
Online exclusive
745.00 
Save 74.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12135-22-7
IUPAC Namepalladium(2+) dihydroxide
Molecular FormulaH2O2Pd
InChI KeyNXJCBFBQEVOTOW-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMoisture: 40.00-65.00 wt%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only