Thermo Scientific Chemicals

Amberlite™ IRN-78, ion exchange resin, nuclear grade, Thermo Scientific Chemicals

Catalog number: 045591.A3
2 kg, Each
Thermo Scientific Chemicals

Amberlite™ IRN-78, ion exchange resin, nuclear grade, Thermo Scientific Chemicals

Catalog number: 045591.A3
2 kg, Each
Quantity
Catalog number: 045591.A3
also known as 045591-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11128-95-3
MDL Number
MFCD00145822
Form
Spherical beads
Moisture holding capacity
≥54.0 to ≤60.0% (as Hydroxide form)
Exchange capacity
≥1.1 eq/liter (as Hydroxide form)
Appearance (Color)
White to yellow to amber
Comment
Coefficient of Uniformity: ≤1.20

Description

Amberlite™ IRN-78 is widely employed in removing radioactive anions for example 131 Iodine and 133 Iodine from waste systems. It also removes the traces of chloride contamination from reactor coolant systems and controls the boron level in the primary system. At higher pH conditions, it is used to remove anionic impurities from secondary streams in the presence of ammonia, morpholine or other amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text