Thermo Scientific Chemicals

Titanium(IV) oxide, anatase, nanopowder, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 045603.36
500 g, Each
Thermo Scientific Chemicals

Titanium(IV) oxide, anatase, nanopowder, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 045603.36
500 g, Each
Quantity
Catalog number: 045603.36
also known as 045603-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-70-0
IUPAC Name
dioxotitanium
Molecular Formula
O2Ti
InChI Key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES
O=[Ti]=O
Comment
15 nm APS
Appearance (Color)
White to pale grey
Total Metal Impurities
≤0.3%
Form
Powder

Description

Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text