m-Carborane, 98%, Thermo Scientific Chemicals
m-Carborane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Carborane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045606.04
Price (USD)/ Each
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS16986-24-6
Molecular FormulaC2H12B10
Molecular Weight (g/mol)144.25
MDL NumberMFCD00151574
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder
Assay by NMR> 97.5% ( by 11B NMR)
Assay by NMR> 97.5% ( by 1H NMR)
m-Carborane is used as a pharmaceutical, chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only