Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%, Thermo Scientific Chemicals
Hafnium(IV) chloride, 99.9% (metals basis), Zr&lt;0.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045612.36
also known as 045612-36
Price (USD)/ Each
1,260.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1483.00 
Save 222.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,260.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1483.00 
Save 222.35 (15%)
Add to cart
Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%, Thermo Scientific Chemicals
Catalog number045612.36
Price (USD)/ Each
1,260.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1483.00 
Save 222.35 (15%)
-
Add to cart
Chemical Identifiers
CAS13499-05-3
IUPAC Namehafnium(4+) tetrachloride
Molecular FormulaCl4Hf
InChI KeyPDPJQWYGJJBYLF-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Hf+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max., Zr<0.5%
Hafnium(IV) chloride is used as a precursor for catalyst involved in the Ziegler-Natta polymerization of propylene. It is an effective Lewis acid for various applications in organic synthesis viz alkylation of ferrocene. Further, it increases the rate and control of 1,3-dipolar cycloadditions. It is also used to prepare hafnium dioxide and hafnium silicate, which is used as high-k-dielectrics in the production of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only