Thermo Scientific Chemicals
ICP-MS Tuning Standard solution for 200.8, Specpure™, Thermo Scientific Chemicals
Catalog number: 045641.AE
100 mL, Each
Thermo Scientific Chemicals
ICP-MS Tuning Standard solution for 200.8, Specpure™, Thermo Scientific Chemicals
Catalog number: 045641.AE
100 mL, Each
Quantity
Catalog number: 045641.AE
also known as 045641-AE
Price (USD)
Have Questions?
Concentration
Be 10?g/mL ?2%
Concentration
Co 10?g/mL ?2%
Concentration
In 10?g/mL ?2%
Concentration
Mg 10?g/mL ?2%
Concentration
Pb 10?g/mL ?2%