Nitinol foil, 0.05mm (0.002in) thick, shape memory, flat annealed, pickled surface, Thermo Scientific Chemicals
Nitinol foil, 0.05mm (0.002in) thick, shape memory, flat annealed, pickled surface, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitinol foil, 0.05mm (0.002in) thick, shape memory, flat annealed, pickled surface, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number 045663.GE
also known as 045663-GE
Price (USD)/ Each
943.00
-
Add to cart
Quantity:
75 x 100 mm
Request bulk or custom format
Price (USD)/ Each
943.00
Add to cart
Nitinol foil, 0.05mm (0.002in) thick, shape memory, flat annealed, pickled surface, Thermo Scientific Chemicals
Catalog number045663.GE
Price (USD)/ Each
943.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialNitinol foil
Name Note0.05mm (0.002 in.) thick, shape memory, flat annealed, pickled surface
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Nitinol foil is used in cell phone antennas, children's toys, experimental materials, eye glasses components and medical equipment. It is also used in heat engines, resilient glass frames, mechanical watch springs, temperature control system, couplings, golf club insert, biomedical and medical uses, cell-phone technology: retractable antenna, microphone boom. It is highly biocompatible and has properties suitable for use in orthopedic implants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only