Thermo Scientific Chemicals

Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals

Catalog number: 045782.18
50 g, Each
Thermo Scientific Chemicals

Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals

Catalog number: 045782.18
50 g, Each
Quantity
Catalog number: 045782.18
also known as 045782-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25655-41-8
Molecular Formula
(C6H9NO)nI2
SMILES
II.*-CC(-*)N1CCCC1=O
MDL Number
MFCD00084483
Appearance (Color)
Yellow to brown
Assay (unspecified)
%Nitrogen: 9.5 to 11.5% ( on a dried basis-
Loss on Drying
8.0% max
Form
powder
Assay (Titration ex Iodide)
% Iodide: 6.6%, max

Description

Polyvinylpyrrolidone-iodine complex is used as a slow iodine release formulation that iodinates lipids and oxidizes cell compounds such as tyrosine side chains. It is utilized to study microbial decontamination of human donor eyes as well as inactivation of infectious hematopoietic necrosis virus. It is also used as a carbon nanotube coating agent to enhance water solubility. It serves as a topical antiseptic in surgery, as an aerosol for skin and mucous membrane infections and is used to prevent infection in wounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text