Thermo Scientific Chemicals

Bis(tricyclohexylphosphine)palladium(0), 97% min, Thermo Scientific Chemicals

Catalog number: 045792.03
1 g, Each
Thermo Scientific Chemicals

Bis(tricyclohexylphosphine)palladium(0), 97% min, Thermo Scientific Chemicals

Catalog number: 045792.03
1 g, Each
Quantity
Catalog number: 045792.03
also known as 045792-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33309-88-5
IUPAC Name
bis(tricyclohexylphosphane) palladium
Molecular Formula
C36H66P2Pd
InChI Key
JGBZTJWQMWZVNX-UHFFFAOYSA-N
SMILES
[Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Form
Crystalline
Assay (unspecified)
97% min
Appearance (Color)
White to tan

Description

Bis(tricyclohexylphosphine)palladium(0) is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text