Silica gel, HPLC/UHPLC grade, spherical, 1.6 micron APS, 110 angstroms, 99.99+%, Thermo Scientific Chemicals
Silica gel, HPLC/UHPLC grade, spherical, 1.6 micron APS, 110 angstroms, 99.99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silica gel, HPLC/UHPLC grade, spherical, 1.6 micron APS, 110 angstroms, 99.99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 045795.06
also known as 045795-06
Price (USD)/ Each
438.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
438.00
Add to cart
Silica gel, HPLC/UHPLC grade, spherical, 1.6 micron APS, 110 angstroms, 99.99+%, Thermo Scientific Chemicals
Catalog number045795.06
Price (USD)/ Each
438.00
-
Add to cart
Specifications
Chemical Name or MaterialSilica gel
Physical FormSpherical, S.A. 340m
Name NoteP.V. 0.95cc/g
CAS63231-67-4
Health Hazard 1Warning
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only