Thermo Scientific Chemicals

Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 045806.30
250 g, Each
Thermo Scientific Chemicals

Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 045806.30
250 g, Each
Quantity
Catalog number: 045806.30
also known as 045806-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1308-06-1
IUPAC Name
dicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular Formula
Co3O4
InChI Key
UBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
Total Metal Impurities
0.3% max.

Description

Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text