Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis), Thermo Scientific Chemicals
Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis), Thermo Scientific Chemicals

May be solid or liquid due to ambient temperature
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 045841.36
also known as 045841-36
Price (USD)/ Each
1,438.65
Online exclusive
1598.00 
Save 159.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,438.65
Online exclusive
1598.00 
Save 159.35 (10%)
Add to cart
Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number045841.36
Price (USD)/ Each
1,438.65
Online exclusive
1598.00 
Save 159.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-55-3
IUPAC Namegallium
Molecular FormulaGa
InChI KeyGYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES[Ga]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid or liquid
Total Metal Impurities≤0.1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only