Thermo Scientific Chemicals

Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v), Thermo Scientific Chemicals

Catalog number: 045844.K2
1 L, Each
Thermo Scientific Chemicals

Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v), Thermo Scientific Chemicals

Catalog number: 045844.K2
1 L, Each
Quantity
Catalog number: 045844.K2
also known as 045844-K2
Price (USD)
Have Questions?

Chemical Identifiers

CAS
64-17-5
IUPAC Name
ethanol
Molecular Formula
C2H6O
InChI Key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES
CCO
Form
Liquid
Assay from Supplier's CofA
Ethyl Acetate: 0.91-1.01%
Appearance (Color)
Clear
Assay from Supplier's CofA
EtOH: 91.12-92.04%
Assay from Supplier's CofA
Toluene: 0.91-1.01%

Description

Denatured ethanol is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text