Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v), Thermo Scientific Chemicals
Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045844.M1
also known as 045844-M1
Price (USD)/ Each
-
Quantity:
4 x 1 L
Request bulk or custom format
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofAEthyl Acetate: 0.91-1.01%
Appearance (Color)Clear
Assay from Supplier's CofAEtOH: 91.12-92.04%
Assay from Supplier's CofAToluene: 0.91-1.01%
View more
Denatured ethanol is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only