Hydroxypropyl methylcellulose, 28-30% methoxyl, 7-12% hydroxypropyl, viscosity (2% aq. soln., 20°C) 7500-14000 mPa.s, Thermo Scientific Chemicals
Hydroxypropyl methylcellulose, 28-30% methoxyl, 7-12% hydroxypropyl, viscosity (2% aq. soln., 20°C) 7500-14000 mPa.s, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydroxypropyl methylcellulose, 28-30% methoxyl, 7-12% hydroxypropyl, viscosity (2% aq. soln., 20°C) 7500-14000 mPa.s, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number 045847.36
also known as 045847-36
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
Add to cart
Hydroxypropyl methylcellulose, 28-30% methoxyl, 7-12% hydroxypropyl, viscosity (2% aq. soln., 20°C) 7500-14000 mPa.s, Thermo Scientific Chemicals
Catalog number045847.36
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9004-65-3
IUPAC Name1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC56H108O30
InChI KeyPUSNGFYSTWMJSK-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤5.0%
pH5.0-8.0
Sulfated Ash≤1.5%
Viscosity7500-14000 mPa.s (2% aq. soln., 20°C)
FormPowder
View more
Hydroxypropyl methylcellulose is used as an ophthalmic lubricant, an emulsifier and a thickening and suspending agent. It is widely used as an excipient in pharmaceutical formulations. It also acts as a food additive. Its eye drops are known as artificial tears, which are used to relieve eye dryness and soreness. It finds applications in various fields as tile adhesives, gypsum products, cosmetics, cement renders, contact lenses, detergents and cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only