Boron Nitride Rod;Diameter (mm), 6.4;Length (mm), 300, Thermo Scientific Chemicals
Boron Nitride Rod;Diameter (mm), 6.4;Length (mm), 300, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron Nitride Rod;Diameter (mm), 6.4;Length (mm), 300, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 045850.KS
Price (USD)/ Each
304.65
Online exclusive
338.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
304.65
Online exclusive
338.00 
Save 33.35 (10%)
Add to cart
Boron Nitride Rod;Diameter (mm), 6.4;Length (mm), 300, Thermo Scientific Chemicals
Catalog number045850.KS
Price (USD)/ Each
304.65
Online exclusive
338.00 
Save 33.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialBoron Nitride
Melting Point2700°C
CAS10043-11-5
Health Hazard 1H335
Health Hazard 3P261-P271-P304+P340-P312-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only