Zeolite Y, hydrogen, Thermo Scientific Chemicals
Zeolite Y, hydrogen, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zeolite Y, hydrogen, Thermo Scientific Chemicals

Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 045868.22
also known as 045868-22
Price (USD)/ Each
83.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
83.00
Add to cart
Zeolite Y, hydrogen, Thermo Scientific Chemicals
Catalog number045868.22
Price (USD)/ Each
83.00
-
Add to cart
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSiO₂:Al₂O₃ mole ratio 5.1:1
CommentSurface area 660 m²/g
Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only