Thermo Scientific Chemicals

Zeolite Y, hydrogen, Thermo Scientific Chemicals

Catalog number: 045871.36
500 g, Each
Thermo Scientific Chemicals

Zeolite Y, hydrogen, Thermo Scientific Chemicals

Catalog number: 045871.36
500 g, Each
Quantity
Catalog number: 045871.36
also known as 045871-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1318-02-1
IUPAC Name
oxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular Formula
Al2O5Si
InChI Key
HNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILES
O=[Si]=O.O=[Al]O[Al]=O
Comment
Surface area 720 m²/g
Comment
SiO₂:Al₂O₃ mole ratio 60:1

Description

Zeolite Y is a useful metal aluminum silicate for proteomics research. It is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. It is also used as a molecular sieve and as an adsorbent of a wide variety of cations, such as Na+, K+, Ca2+, Mg2+ and others. Zeolite is used in the development of a variety of bioreactor devices and in the determination of serum triglycerides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text