Zeolite beta, ammonium, Thermo Scientific Chemicals
Zeolite beta, ammonium, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zeolite beta, ammonium, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 045873.36
also known as 045873-36
Price (USD)/ Each
366.65
Online exclusive
407.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
366.65
Online exclusive
407.00 
Save 40.35 (10%)
Add to cart
Zeolite beta, ammonium, Thermo Scientific Chemicals
Catalog number045873.36
Price (USD)/ Each
366.65
Online exclusive
407.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSurface area 680 m²/g
CommentSiO₂:Al₂O₃ mole ratio 25:1
Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
RUO – Research Use Only