Thermo Scientific Chemicals

Zeolite beta, ammonium, Thermo Scientific Chemicals

Catalog number: 045874.14
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Zeolite beta, ammonium, Thermo Scientific Chemicals

Catalog number: 045874.14
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 045874.14
also known as 045874-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1318-02-1
IUPAC Name
oxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular Formula
Al2O5Si
InChI Key
HNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILES
O=[Si]=O.O=[Al]O[Al]=O
Comment
Surface area 710 m²/g
Comment
SiO₂:Al₂O₃ mole ratio 38:1

Description

Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text