Thermo Scientific Chemicals

Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals

Catalog number: 045879.36
500 g, Each
Thermo Scientific Chemicals

Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals

Catalog number: 045879.36
500 g, Each
Quantity
Catalog number: 045879.36
also known as 045879-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1318-02-1
IUPAC Name
oxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular Formula
Al2O5Si
InChI Key
HNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILES
O=[Si]=O.O=[Al]O[Al]=O
Comment
SiO₂:Al₂O₃ mole ratio 23:1
Comment
Surface area 425 m²/g

Description

Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst supportZeolite ZSM-5 is used in hydrocarbon isomerization, alkylation of hydrocarbons. It is also used as a catalyst support. For example, copper is deposited on the zeolite and utilized for the preparation of acetaldehyde from ethanol by oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text