Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 045881.36
also known as 045881-36
Price (USD)/ Each
348.65
Online exclusive
387.00 
Save 38.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
348.65
Online exclusive
387.00 
Save 38.35 (10%)
Add to cart
Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
Catalog number045881.36
Price (USD)/ Each
348.65
Online exclusive
387.00 
Save 38.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSurface area 425 m²/g
CommentSiO₂:Al₂O₃ mole ratio 50:1
Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only