Thermo Scientific Chemicals

Zeolite ferrierite, ammonium, Thermo Scientific Chemicals

Catalog number: 045884.22
100 g, Each
Thermo Scientific Chemicals

Zeolite ferrierite, ammonium, Thermo Scientific Chemicals

Catalog number: 045884.22
100 g, Each
Quantity
Catalog number: 045884.22
also known as 045884-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1318-02-1
IUPAC Name
oxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular Formula
Al2O5Si
InChI Key
HNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILES
O=[Si]=O.O=[Al]O[Al]=O
Comment
Surface area 400 m²/g
Comment
SiO₂:Al₂O₃ mole ratio 20:1

Description

Zeolite ferrierite, ammonium is used as a commercial filter and ion-exchange bed. Further, it acts as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite ferrierite is used as commercial filters, ion-exchange beds, and as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text