Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals
Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 045888.22
also known as 045888-22
Price (USD)/ Each
24.65
Online exclusive
27.50 
Save 2.85 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
24.65
Online exclusive
27.50 
Save 2.85 (10%)
Add to cart
Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals
Catalog number045888.22
Price (USD)/ Each
24.65
Online exclusive
27.50 
Save 2.85 (10%)
-
Add to cart
Chemical Identifiers
CAS22326-55-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Barium hydroxide monohydrate is used in water purification. It is used to prepare lubricating and oil additives as well as a precursor to other barium compounds. It is also used to dehydrate and remove sulfate from various products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only