Thermo Scientific Chemicals

Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 045888.36
500 g, Each
Thermo Scientific Chemicals

Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 045888.36
500 g, Each
Quantity
Catalog number: 045888.36
also known as 045888-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22326-55-2
Assay (unspecified)
94.1% min

Description

Barium hydroxide monohydrate is used in water purification. It is used to prepare lubricating and oil additives as well as a precursor to other barium compounds. It is also used to dehydrate and remove sulfate from various products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text