Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals
Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number 045896.36
Price (USD)/ Each
211.65
Online exclusive
235.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
211.65
Online exclusive
235.00 
Save 23.35 (10%)
Add to cart
Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals
Catalog number045896.36
Price (USD)/ Each
211.65
Online exclusive
235.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS68083-14-7
IUPAC Name3,3-dimethyl-5,5-diphenyl-2,4-dioxa-3,5-disilahexane
Molecular FormulaC16H22O2Si2
InChI KeyARWRSWALIGRKQA-UHFFFAOYSA-N
SMILESCO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Formclear, viscous liquid
Refractive Index1.500 to 1.510 ( at 25 deg C)
Appearance (Color)Colorless
Assay (unspecified)% volatile ( 24 hours, @ 150 deg C):
Silicone oil is used in high temperature open systems, closed systems and circulating, closed loop and heat transfer baths. It is used as a high temperature fluid for laboratory research apparatus and instruments. It is also used as a calibration media in temperature controlled baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High temperature open system, closed system, circulating, closed loop, and heat transfer baths
  • High temperature fluids for laboratory research apparatus and instruments
RUO – Research Use Only