Thermo Scientific Chemicals

Aluminum oxide, super activated, neutral, Grade I, Thermo Scientific Chemicals

Catalog number: 045901.A1
1 kg, Each
Thermo Scientific Chemicals

Aluminum oxide, super activated, neutral, Grade I, Thermo Scientific Chemicals

Catalog number: 045901.A1
1 kg, Each
Quantity
Catalog number: 045901.A1
also known as 045901-A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Particle Size
50-200 Microns

Description

Aluminum oxide is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories. Used in protein extraction as a component in the preparation of tissues. Used in bushings, spindles, lapping, polishing, refractory coating for investment casting shells, decorative non-skid tile, and pressure blasting. It is also used in varnishes, for textile impregnation, in cosmetics and as an intermediate in pharmaceutical production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text