Poly(3-hexylthiophene-2,5-diyl), regiorandom, Thermo Scientific Chemicals
Poly(3-hexylthiophene-2,5-diyl), regiorandom, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(3-hexylthiophene-2,5-diyl), regiorandom, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045906.ME
also known as 045906-ME
Price (USD)/ Each
-
Quantity:
500 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly (3-hexylthiophene-2,5-diyl)
ColorBrown
CAS104934-50-1
Physical FormPowder/Chunks
Health Hazard 1H315-H319-H335
View more
Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) research and devices. It is also used in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only