Boron Nitride rod is used as a container for most molten metals like aluminum, iron, steel, silicon, tin germanium and copper due to its properties like corrosion resistance, electrical resistance and the fact that it can withstand temperatures of about 2000
0. It plays an important role in nanotechnology to produce nanotubes, which find applications in the aerospace industry. It is widely utilized in evaporator boats, nozzles for nonferrous metals, electric insulators in vacuum system, induction heating coil supports, crucibles and radar components and antenna windows and insulators for high temperature furnace.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.