Palladium, 5% on activated carbon paste, 5R394, Thermo Scientific Chemicals
Palladium, 5% on activated carbon paste, 5R394, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 5% on activated carbon paste, 5R394, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045921.06
also known as 045921-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium
Name Note5% on activated carbon paste, 5R394
CAS7440-05-3
ColorBlack
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only