Thermo Scientific Chemicals

Antimony(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 045922.09
10 g, Each
Thermo Scientific Chemicals

Antimony(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 045922.09
10 g, Each
Quantity
Catalog number: 045922.09
also known as 045922-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1327-50-0
IUPAC Name
bis(tellanylidenestibanyl) tellurium
Molecular Formula
Sb2Te3
InChI Key
BPDQXJZWVBPDSN-UHFFFAOYSA-N
SMILES
[Te].[Sb]=[Te].[Sb]=[Te]
Total Metal Impurities
0.001% max.

Description

Antimony(III) telluride is used as both n-type and p-type semiconductors in association with appropriate dopant. It reacts with germanium telluride and forms the pseudobinary intermetallic system. It is used in solid state refrigerators due to its large thermoelectric effect. Further, it is an active component of thermoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text