Thermo Scientific Chemicals

Lithium perchlorate trihydrate, ACS, 63.0-68.0% LiClO{4}, Thermo Scientific Chemicals

Catalog number: 045923.22
100 g, Each
Thermo Scientific Chemicals

Lithium perchlorate trihydrate, ACS, 63.0-68.0% LiClO{4}, Thermo Scientific Chemicals

Catalog number: 045923.22
100 g, Each
Quantity
Catalog number: 045923.22
also known as 045923-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-78-6
IUPAC Name
lithium(1+) trihydrate perchlorate
Molecular Formula
ClH6LiO7
InChI Key
JAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES
[Li+].O.O.O.[O-][Cl](=O)(=O)=O
Heavy Metals
(ICP-OES) 5 ppm
Impurity content
Cl 0.003%
Impurity content
SO₄ 0.001%
pH
6.0-7.5
Assay (unspecified)
63.0-68.0% LiClO_4

Description

Lithium perchlorate trihydrate is used as an oxygen source for chemical oxygen generators as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte and acts as a chaotropic agent to denature proteins. It is used in the Baylis-Hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text