Thermo Scientific Chemicals

Aluminum oxide, neutral, HPLC Flash Grade, Thermo Scientific Chemicals

Catalog number: 046025.A1
1 kg, Each
Thermo Scientific Chemicals

Aluminum oxide, neutral, HPLC Flash Grade, Thermo Scientific Chemicals

Catalog number: 046025.A1
1 kg, Each
Quantity
Catalog number: 046025.A1
also known as 046025-A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
pH
of a 5% slurry 6.5-7.5
Particle Size
18-32 Microns

Description

Aluminum oxide is an inorganic compound used as a grinding or blending agent. In addition, Aluminum oxide can be essential in protein extraction as a component in the preparation of tissues. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text