Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Type | Eluent Concentrate |
Standard Concentration | 5 X |
Hazards | Irritant |
Standard Components | Pyridine-2,6-dicarboxylic Acid, Potassium Hydroxide, Potassium Sulfate, Formic Acid in Deionized Water |
For Use With (Application) | Ion Chromatography |
Volume (Metric) | 1000 mL |
Description | MetPac PDCA Eluent Concentrate (5 x); 1000 mL |
Storage | Room Temperature |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Volume (Metric) | For Use With (Application) | Price (USD) | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
046088 | Each 1 | 1000 mL | Ion Chromatography | 142.00 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The PDCA Eluent Concentrate, is available in 1000mL and contains the following;
The Dionex PDCA transition metals analysis reagent is a chelating agent for transition metals eluents, available in 20g and contains the following;
Only high-purity, reagent-grade chemicals and water (18MO-cm), quality-control checked for ionic contamination, are used in Thermo Scientific Dionex reagents, standards and eluent concentrates.