Magnesium thiosulfate hexahydrate, 99%, Thermo Scientific Chemicals
Magnesium thiosulfate hexahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium thiosulfate hexahydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 046114.22
also known as 046114-22
Price (USD)/ Each
46.60
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
46.60
Add to cart
Magnesium thiosulfate hexahydrate, 99%, Thermo Scientific Chemicals
Catalog number046114.22
Price (USD)/ Each
46.60
-
Add to cart
Chemical Identifiers
CAS13446-30-5
IUPAC Namemagnesium(2+) hexahydrate sulfanidesulfonate
Molecular FormulaH12MgO9S2
InChI KeyCQDMJJVHDPDPHO-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Magnesium hyposulfite finds application in the treatment of certain skin diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only