Thermo Scientific Chemicals

Magnesium thiosulfate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 046114.36
500 g, Each
Thermo Scientific Chemicals

Magnesium thiosulfate hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 046114.36
500 g, Each
Quantity
Catalog number: 046114.36
also known as 046114-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13446-30-5
IUPAC Name
magnesium(2+) hexahydrate sulfanidesulfonate
Molecular Formula
H12MgO9S2
InChI Key
CQDMJJVHDPDPHO-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Mg++].[O-]S([S-])(=O)=O
Assay (unspecified)
98.5% min

Description

Magnesium hyposulfite finds application in the treatment of certain skin diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text