Thermo Scientific Chemicals

Magnesium bromide hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 046120.22
100 g, Each
Thermo Scientific Chemicals

Magnesium bromide hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 046120.22
100 g, Each
Quantity
Catalog number: 046120.22
also known as 046120-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13446-53-2
IUPAC Name
dibromomagnesium hexahydrate
Molecular Formula
Br2H12MgO6
InChI Key
LGLXXNHIGIJYQQ-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.Br[Mg]Br
Assay (unspecified)
98.1% min

Description

Magnesium bromide hexahydrate is used as flame retardant. Used as source of magnesium ion and a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text