Aluminum oxide, Drysphere∣r desiccant, without indicator, Thermo Scientific Chemicals
Aluminum oxide, Drysphere∣r desiccant, without indicator, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, Drysphere∣r desiccant, without indicator, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Catalog number 046172.22
also known as 046172-22
Price (USD)/ Each
29.60
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
29.60
Add to cart
Aluminum oxide, Drysphere∣r desiccant, without indicator, Thermo Scientific Chemicals
Catalog number046172.22
Price (USD)/ Each
29.60
-
Add to cart
Specifications
Boiling Point2980°C
Chemical Name or MaterialAluminum oxide, Drysphere™ desiccant
Melting Point2045°C
Name NoteWithout indicator
CAS1344-28-1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only