Thermo Scientific Chemicals

Buffer solution, pH 13.00 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Catalog number: 046307.K7
4 L, Each
Thermo Scientific Chemicals

Buffer solution, pH 13.00 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Catalog number: 046307.K7
4 L, Each
Quantity
Catalog number: 046307.K7
also known as 046307-K7
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Clear colorless
pH
13.00 (+/-0.026 @ 25°C)
Form
Liquid

Description

Buffer solution, pH 13.00 (±0.01 at 25°C), No Color, Specpure™, NIST Traceable is used as a means of keeping the pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text