Thermo Scientific Chemicals

Fullerene powder, 99% C{6}{0}, Thermo Scientific Chemicals

Catalog number: 046311.06
5 g, Each
Thermo Scientific Chemicals

Fullerene powder, 99% C{6}{0}, Thermo Scientific Chemicals

Catalog number: 046311.06
5 g, Each
Quantity
Catalog number: 046311.06
also known as 046311-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99685-96-8
IUPAC Name
(C60-Ih)[5,6]fullerene
Molecular Formula
C60
InChI Key
XMWRBQBLMFGWIX-UHFFFAOYSA-N
SMILES
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Appearance (Color)
Black
Form
powder
Assay (HPLC)
> 98.5%

Description

It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text