Thermo Scientific Chemicals

Tetra-n-butylammonium hydroxide, 1.0M aq. soln., Thermo Scientific Chemicals

Catalog number: 046315.AE
100 mL, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium hydroxide, 1.0M aq. soln., Thermo Scientific Chemicals

Catalog number: 046315.AE
100 mL, Each
Quantity
Catalog number: 046315.AE
also known as 046315-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2052-49-5
IUPAC Name
tetrabutylazanium hydroxide
Molecular Formula
C16H37NO
InChI Key
VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Color)
Clear, almost colorless liquid
Specific Absorbance (E 1%/1cm)
254nm ≤0.10 AU (measured at 0.1 M concentration)
Specific Absorbance (E 1%/1cm)
240nm ≤0.15 AU (measured at 0.1 M concentration)
Specific Absorbance (E 1%/1cm)
300nm ≤0.05 AU (measured at 0.1 M concentration)
Form
Liquid

Description

Tetra-n-butylammonium hydroxide is used in biological studies for the detection of counter-anion effect on antimicrobial activity of tetrabutylammonium salts, intermediates, Plating agents and surface treating agents, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text