Thermo Scientific Chemicals

Copper(II) ethoxide, 98%, Thermo Scientific Chemicals

Catalog number: 046316.06
5 g, Each
Thermo Scientific Chemicals

Copper(II) ethoxide, 98%, Thermo Scientific Chemicals

Catalog number: 046316.06
5 g, Each
Quantity
Catalog number: 046316.06
also known as 046316-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2850-65-9
IUPAC Name
copper(2+) diethanolate
Molecular Formula
C4H10CuO2
InChI Key
CRCKGIUJMFFISH-UHFFFAOYSA-N
SMILES
[Cu++].CC[O-].CC[O-]
Appearance (Color)
Blue to blue-green to green
Assay from Supplier's CofA
≥97.5%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text