Thermo Scientific Chemicals

Ammonium pentaborate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 046323.A3
2 kg, Each
Thermo Scientific Chemicals

Ammonium pentaborate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 046323.A3
2 kg, Each
Quantity
Catalog number: 046323.A3
also known as 046323-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12046-04-7
IUPAC Name
nonaammonium tetrahydrate pentaborate
Molecular Formula
B5H44N9O19
InChI Key
OJAJAQRDJBKPPU-UHFFFAOYSA-W
SMILES
[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O.O.O.O.[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-].[O-]B([O-])[O-]
Form
Powder
Appearance (Color)
White
Assay (unspecified)
63% B2O3 min

Description

Ammonium pentaborate is used in the preparation of both wet and dry electrolytic capacitors, flame proofing, gunning and patching compounds for extending the life of basic refractories. Used in the preparation of special welding, soldering and brazing fluxes for stainless steel or various non-ferrous metals, where alkali borates cannot be used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text