Potassium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
4 L
Catalog number 046330.K2
also known as 046330-K2
Price (USD)/ Each
54.65
Online exclusive
60.50 
Save 5.85 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
60.50 
Save 5.85 (10%)
Add to cart
Potassium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Catalog number046330.K2
Price (USD)/ Each
54.65
Online exclusive
60.50 
Save 5.85 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥23.75 to ≤26.25%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only