Thermo Scientific Chemicals

6-Maleimidohexanoic acid, Thermo Scientific Chemicals

Catalog number: 046384.14
25 g, Each
Thermo Scientific Chemicals

6-Maleimidohexanoic acid, Thermo Scientific Chemicals

Catalog number: 046384.14
25 g, Each
Quantity
Catalog number: 046384.14
also known as 046384-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55750-53-3
IUPAC Name
6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoic acid
Molecular Formula
C10H13NO4
InChI Key
WOJKKJKETHYEAC-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCN1C(=O)C=CC1=O
Assay (unspecified)
Wt loss at 100 deg C: < 1.0 wt%
Assay (unspecified)
Wt loss at 300 deg C: < 35.0 wt%
Form
powder
Appearance (Color)
Pale yellow
Assay (unspecified)
> 97.0% (Chromatography)

Description

6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text